Tisk…

2-Chloro-5-nitrocinnamic acid 97%

Dodavatel: Apollo Scientific

OR2084-25G OR2084-5G
APOSOR2084-25GEA 4100 CZK
APOSOR2084-25G APOSOR2084-5G
2-Chloro-5-nitrocinnamic acid 97%
2-Chloro-5-nitrocinnamic acid
Vzorec: C₉H₆ClNO₄
Bod tání: 219…221 °C
Storage Temperature: Ambient
MDL Number: MFCD00063312
CAS číslo: 36015-19-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR