Tisk…

Methyl-3-amino-4-chlorobenzoate

Dodavatel: Apollo Scientific
Varování

Synonyms: Methyl 3-amino-4-chlorobenzoate, Methyl 3-Amino-4-chlorobenzoate, 3-amino-4-chlorobenzoic acid methylester

OR2041-100G OR2041-25G OR2041-5G
APOSOR2041-100GEA 2980 CZK
APOSOR2041-100G APOSOR2041-25G APOSOR2041-5G
Methyl-3-amino-4-chlorobenzoate
Methyl 3-amino-4-chlorobenzoate
Vzorec: C₈H₈ClNO₂
MW: 185,6100 g/mol
Bod tání: 82…83 °C
Storage Temperature: Ambient
MDL Number: MFCD03470533
CAS číslo: 40872-87-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR