Tisk…

3-Bromopyridine-2-carbonitrile 97+%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 3-Bromopicolinonitrile, 3-Bromo-2-cyanopyridine

OR2027-1G OR2027-25G OR2027-5G
APOSOR2027-1GEA 622 CZK
APOSOR2027-1G APOSOR2027-25G APOSOR2027-5G
3-Bromopyridine-2-carbonitrile 97+%
3-Bromopyridine-2-carbonitrile
Vzorec: C₆H₃BrN₂
MW: 183.01
Bod varu: 120 °C/4mmHg
Bod tání: 93…99 °C
Storage Temperature: Ambient
MDL Number: MFCD02683288
CAS číslo: 55758-02-6
UN: 2811
ADR: 6.1 III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR