5-Methylfuran-2-boronic acid 97%
Dodavatel: Apollo Scientific
|
Varování
|
OR2011-1G
OR2011-250MG
OR2011-5G
APOSOR2011-1GEA
1580
CZK
APOSOR2011-1G
APOSOR2011-250MG
APOSOR2011-5G
5-Methylfuran-2-boronic acid 97%
5-Methylfuran-2-boronic acid
Vzorec:
C₅H₇BO₃ Bod tání: 82 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD01114644 CAS číslo: 62306-79-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...