Tisk…

Trimethylsulphoxonium chloride

Dodavatel: Apollo Scientific
Varování

Synonyms: Trimethyloxosulphonium chloride

OR20031-100G OR20031-25G OR20031-5G
APOSOR20031-100GEA 2400 CZK
APOSOR20031-100G APOSOR20031-25G APOSOR20031-5G
Trimethylsulphoxonium chloride
Trimethylsulphoxonium chloride
Vzorec: C₃H₉ClOS
MW: 128,62 g/mol
Bod tání: 226…229 °C
Storage Temperature: Ambient
MDL Number: MFCD00011898
CAS číslo: 5034-06-0
EINECS: 225-724-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR