2-Bromopyridine-6-carbonyl chloride
Dodavatel: Apollo Scientific
OR200166-1G
OR200166-50MG
OR200166-250MG
OR200166-500MG
APOSOR200166-1GEA
13630
CZK
APOSOR200166-1G
APOSOR200166-50MG
APOSOR200166-250MG
APOSOR200166-500MG
2-Bromopyridine-6-carbonyl chloride
2-Bromopyridine-6-carbonyl chloride
Vzorec:
C₆H₃BrClNO Storage Temperature: Ambient |
MDL Number:
MFCD18909527 CAS číslo: 139487-69-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...