Tisk…

Propionamidine hydrochloride 97%

Dodavatel: Apollo Scientific
Varování

Synonyms: Propionimidamide hydrochloride

OR200136-1G OR200136-10G OR200136-5G
APOSOR200136-1GEA 622 CZK
APOSOR200136-1G APOSOR200136-10G APOSOR200136-5G
Propionamidine hydrochloride 97%
Propionamidine hydrochloride
Vzorec: C₃H₉ClN₂
MW: 108.569 g/mol
Bod tání: 128…130 °C
Storage Temperature: Ambient
MDL Number: MFCD00233938
CAS číslo: 3599-89-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR