2-Methyl-1,3-oxazole-4-carbonyl chloride
Dodavatel: Apollo Scientific
|
Nebezpečí
|
OR200129-1G
OR200129-250MG
OR200129-100MG
OR200129-500MG
APOSOR200129-1GEA
14150
CZK
APOSOR200129-1G
APOSOR200129-250MG
APOSOR200129-100MG
APOSOR200129-500MG
2-Methyl-1,3-oxazole-4-carbonyl chloride
2-Methyl-1,3-oxazole-4-carbonyl chloride
Vzorec:
C₅H₄ClNO₂ Storage Temperature: Ambient |
MDL Number:
MFCD13173537 CAS číslo: 62348-22-5 UN: 1760 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...