Tisk…

3,5-Dichloropyrazine-2-carbaldehyde

Dodavatel: Apollo Scientific
Varování

OR20012-1G OR20012-50MG OR20012-250MG OR20012-500MG
APOSOR20012-1GEA 29630 CZK
APOSOR20012-1G APOSOR20012-50MG APOSOR20012-250MG APOSOR20012-500MG
3,5-Dichloropyrazine-2-carbaldehyde
3,5-Dichloropyrazine-2-carbaldehyde
Vzorec: C₅H₂Cl₂N₂O
MW: 176,99 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09864908
CAS číslo: 136866-27-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR