Tisk…

5-Formyl-2-furancarboxylic acid

Dodavatel: Apollo Scientific

Synonyms: Furfural-5-carboxylic Acid, 5-Formylfuran-2-carboxylic acid, 5-Formyl-2-furoic Acid

OR200110-250MG OR200110-500MG
APOSOR200110-250MGEA 2700 CZK
APOSOR200110-250MG APOSOR200110-500MG
5-Formyl-2-furancarboxylic acid
5-Formyl-2-furancarboxylic acid
Vzorec: C₆H₄O₄
MW: 140,09 g/mol
Bod tání: 209 °C
Storage Temperature: Ambient
MDL Number: MFCD00020924
CAS číslo: 13529-17-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR