Tisk…

1-[2-(4-Bromophenoxy)ethyl]piperidine

Dodavatel: Apollo Scientific
Varování

OR200097-1G OR200097-250MG OR200097-5G OR200097-500MG
APOSOR200097-1GEA 7340 CZK
APOSOR200097-1G APOSOR200097-250MG APOSOR200097-5G APOSOR200097-500MG
1-[2-(4-Bromophenoxy)ethyl]piperidine
1-[2-(4-Bromophenoxy)ethyl]piperidine
Vzorec: C₁₃H₁₈BrNO
Storage Temperature: Ambient
MDL Number: MFCD01308459
CAS číslo: 836-58-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR