Tisk…

4-Isopropylphenylacetyl chloride

Dodavatel: Apollo Scientific

OR200093-1G OR200093-250MG OR200093-500MG
APOSOR200093-1GEA 12670 CZK
APOSOR200093-1G APOSOR200093-250MG APOSOR200093-500MG
4-Isopropylphenylacetyl chloride
4-Isopropylphenylacetyl chloride
Vzorec: C₁₁H₁₃ClO
Storage Temperature: Ambient
MDL Number: MFCD12024949
CAS číslo: 52629-44-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR