Tisk…

4-sulphamoylbenzoyl chloride

Dodavatel: Apollo Scientific

OR200092-1G OR200092-250MG OR200092-2.5G
APOSOR200092-1GEA 12670 CZK
APOSOR200092-1G APOSOR200092-250MG APOSOR200092-2.5G
4-sulphamoylbenzoyl chloride
4-sulphamoylbenzoyl chloride
Vzorec: C₇H₆ClNO₃S
MW: 219.65 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09755960
CAS číslo: 51594-97-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR