Tisk…

4-Oxo-4-(4-propylphenyl)butanoyl chloride

Dodavatel: Apollo Scientific

OR200086-1G OR200086-250MG OR200086-500MG OR200086-2.5G OR200086-5G
APOSOR200086-1GEA 12300 CZK
APOSOR200086-1G APOSOR200086-250MG APOSOR200086-500MG APOSOR200086-2.5G APOSOR200086-5G
4-Oxo-4-(4-propylphenyl)butanoyl chloride
4-Oxo-4-(4-propylphenyl)butanoyl chloride
Vzorec: C₁₃H₁₅ClO₂
Storage Temperature: Ambient
MDL Number: MFCD23703527
CAS číslo: 1429309-23-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR