Morpholine-4-carboxamidinium acetate
Dodavatel: Apollo Scientific
OR200058-1G
OR200058-5G
APOSOR200058-1GEA
0
CZK
APOSOR200058-1G
APOSOR200058-5G
Morpholine-4-carboxamidinium acetate
Morpholine-4-carboxamidinium acetate
Vzorec:
C₇H₁₅N₃O₃ MW: 189.22 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03428317 CAS číslo: 402726-73-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...