1-(2-Aminoethyl)pyrrolidine 98%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
N-(2-Aminoethyl)pyrrolidine, Pyrrolidinoethylamine
OR19557-100G
OR19557-25G
OR19557-5G
APOSOR19557-100GEA
5890
CZK
APOSOR19557-100G
APOSOR19557-25G
APOSOR19557-5G
1-(2-Aminoethyl)pyrrolidine 98%
1-(2-Aminoethyl)pyrrolidine
Vzorec:
C₆H₁₄N₂ MW: 114,19 g/mol Bod varu: 161 °C Hustota: 0,901 g/cm³ (25 °C) Bod vzplanutí: 47 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003182 CAS číslo: 7154-73-6 EINECS: 230-509-5 UN: 2734 ADR: 8+3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...