Tisk…

(S)-Glycidyl trityl ether 98%

Dodavatel: Apollo Scientific
Varování

Synonyms: (S)-Glycidyl Triphenylmethyl Ether, (S)-2-(Triphenylmethoxymethyl)oxirane, (S)-2-((Trityloxy)methyl)oxirane

OR19556-100G
APOSOR19556-100GEA 2860 CZK
APOSOR19556-100G
(S)-Glycidyl trityl ether 98%
(S)-Glycidyl trityl ether
Vzorec: C₂₂H₂₀O₂
MW: 316,40 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00273373
CAS číslo: 129940-50-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR