4-(Aminoethyl)-1-N-boc-piperidine
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
4-(Aminoethyl)-1-N-Boc-piperidine, 4-(2-Aminoethyl)piperidine, N1-BOC protected, tert-butyl 4-(2-aminoethyl)piperidine-1-carboxylate
OR19534-1G
OR19534-5G
APOSOR19534-1GEA
2240
CZK
APOSOR19534-1G
APOSOR19534-5G
4-(Aminoethyl)-1-N-boc-piperidine
4-(Aminoethyl)-1-N-boc-piperidine
Vzorec:
C₁₂H₂₄N₂O₂ MW: 228,3312 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD04038459 CAS číslo: 146093-46-1 UN: 3259 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...