Tisk…

2-Bromo-4-(tert-butyl)aniline

Dodavatel: Apollo Scientific
Varování

OR19485-25G OR19485-5G
APOSOR19485-25GEA 3190 CZK
APOSOR19485-25G APOSOR19485-5G
2-Bromo-4-(tert-butyl)aniline
2-Bromo-4-(tert-butyl)aniline
Vzorec: C₁₀H₁₄BrN
MW: 228,13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00052693
CAS číslo: 103273-01-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR