5-Bromoisophthalic acid 97%
Dodavatel: Apollo Scientific
|
Varování
|
OR19484-25G
OR19484-100G
APOSOR19484-25GEA
1660
CZK
APOSOR19484-25G
APOSOR19484-100G
5-Bromoisophthalic acid 97%
5-Bromoisophthalic acid
Vzorec:
C₈H₅BrO₄ MW: 245,03 g/mol Bod tání: 276…278 °C Storage Temperature: Ambient |
MDL Number:
MFCD00152019 CAS číslo: 23351-91-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...