Tisk…

4-(Methoxymethyl)phenylacetic acid

Dodavatel: Apollo Scientific
Varování

Synonyms: 2-(4-(Methoxymethyl)phenyl)acetic acid

OR19480-1G OR19480-250MG OR19480-5G
APOSOR19480-1GEA 6850 CZK
APOSOR19480-1G APOSOR19480-250MG APOSOR19480-5G
4-(Methoxymethyl)phenylacetic acid
4-(Methoxymethyl)phenylacetic acid
Vzorec: C₁₀H₁₂O₃
MW: 180,21 g/mol
Bod tání: 47…49 °C
Storage Temperature: Ambient
MDL Number: MFCD08060493
CAS číslo: 343880-24-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR