Tisk…
(2-Nitrophenyl)hydrazine hydrochloride
Dodavatel: Apollo Scientific
Nebezpečí
Synonyms: 2-Nitrophenylhydrazine hydrochloride
OR1939-100G OR1939-25G OR1939-5G
APOSOR1939-100GEA 2360 CZK
APOSOR1939-100G APOSOR1939-25G APOSOR1939-5G
(2-Nitrophenyl)hydrazine hydrochloride
(2-Nitrophenyl)hydrazine hydrochloride
Vzorec: C₆H₈ClN₃O₂
MW: 189,6 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00060205
CAS číslo: 6293-87-4
UN: 1325
ADR: 4.1,II