Tisk…

Cyclopropanemethylamine 97%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: Cyclopropylmethylamine, (Cyclopropylmethyl)amine, (Aminomethyl)cyclopropane

OR1937-25G OR1937-5G
APOSOR1937-25GEA 2280 CZK
APOSOR1937-25G APOSOR1937-5G
Cyclopropanemethylamine 97%
Cyclopropanemethylamine
Vzorec: C₄H₉N
MW: 71,12 g/mol
Bod varu: 84…85 °C
Hustota: 0,82 g/cm³ (25 °C)
Bod vzplanutí: 2 °C
Storage Temperature: Ambient
MDL Number: MFCD00037147
CAS číslo: 2516-47-4
EINECS: 219-737-6
UN: 2733
ADR: 3+8,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR