4-tert-Butylcyclohexene-1-ylboronic acid 96%
Dodavatel: Apollo Scientific
OR1926-1G
OR1926-250MG
APOSOR1926-1GEA
14880
CZK
APOSOR1926-1G
APOSOR1926-250MG
4-tert-Butylcyclohexene-1-ylboronic acid 96%
4-tert-Butylcyclohexene-1-ylboronic acid
Vzorec:
C₁₀H₁₉BO₂ Storage Temperature: Ambient |
MDL Number:
MFCD06659856 CAS číslo: 850567-91-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...