Tisk…

4-Ethoxy-3-methylbenzeneboronic acid 97%

Dodavatel: Apollo Scientific
Varování

Synonyms: 4-Ethoxy-3-methylphenylboronic acid

OR1925-5G OR1925-25G
APOSOR1925-5GEA 2780 CZK
APOSOR1925-5G APOSOR1925-25G
4-Ethoxy-3-methylbenzeneboronic acid 97%
4-Ethoxy-3-methylbenzeneboronic acid
Vzorec: C₉H₁₃BO₃
MW: 180,01 g/mol
Bod tání: 168…172 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06659870
CAS číslo: 850568-08-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR