Tisk…

5-Bromo-2-hydroxybenzeneboronic acid 97%

Dodavatel: Apollo Scientific
Varování

Synonyms: (5-Bromo-2-hydroxyphenyl)boronic acid, 5-Bromo-2-hydroxyphenylboronic acid

OR1920-1G OR1920-5G
APOSOR1920-1GEA 2690 CZK
APOSOR1920-1G APOSOR1920-5G
5-Bromo-2-hydroxybenzeneboronic acid 97%
5-Bromo-2-hydroxybenzeneboronic acid
Vzorec: C₆H₆BBrO₃
MW: 216,83 g/mol
Bod tání: 196…200 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00093408
CAS číslo: 89598-97-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR