Tisk…

6-Aminopicolinic acid 98%

Dodavatel: Apollo Scientific
Varování

OR1917-25G OR1917-5G OR1917-100G
APOSOR1917-25GEA 2980 CZK
APOSOR1917-25G APOSOR1917-5G APOSOR1917-100G
6-Aminopicolinic acid 98%
6-Aminopicolinic acid
Vzorec: C₆H₆N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD00233711
CAS číslo: 23628-31-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR