Tisk…

2,6-Dichloronicotinic acid 97%

Dodavatel: Apollo Scientific
Varování

Synonyms: 2,6-Dichloro-3-pyridinecarboxylic acid, 2,6-Dichloropyridine-3-carboxylic acid

OR1907-100G OR1907-25G OR1907-5G
APOSOR1907-100GEA 3060 CZK
APOSOR1907-100G APOSOR1907-25G APOSOR1907-5G
2,6-Dichloronicotinic acid 97%
2,6-Dichloronicotinic acid
Vzorec: C₆H₃Cl₂NO₂
MW: 192 g/mol
Bod tání: 140…145 °C
Storage Temperature: Ambient
MDL Number: MFCD00075583
CAS číslo: 38496-18-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR