2-Bromoisonicotinic acid 97%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
2-Bromo-4-pyridinecarboxylic acid, 2-Bromopyridine-4-carboxylic acid
OR1905-5G
OR1905-25G
APOSOR1905-5GEA
622
CZK
APOSOR1905-5G
APOSOR1905-25G
2-Bromoisonicotinic acid 97%
2-Bromoisonicotinic acid
Vzorec:
C₆H₄BrNO₂ MW: 202,01 g/mol Bod tání: 217…229 °C Storage Temperature: Ambient |
MDL Number:
MFCD01646069 CAS číslo: 66572-56-3 |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...