Acetoxyacetyl chloride
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
Acetoxyacetylchloride, 2-Chloro-2-oxoethyl acetate
OR1899-100G
OR1899-25G
OR1899-5G
APOSOR1899-100GEA
3810
CZK
APOSOR1899-100G
APOSOR1899-25G
APOSOR1899-5G
Acetoxyacetyl chloride
Acetoxyacetyl chloride
Vzorec:
C₄H₅ClO₃ Bod varu: 55 °C/12mmHg Hustota: 1,27 g/cm³ (25 °C) Bod vzplanutí: 71 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00011535 CAS číslo: 13831-31-7 EINECS: 237-542-4 UN: 3265 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...