4-(Benzyloxy)cyclohexanone 96%
Dodavatel: Apollo Scientific
|
Varování
|
OR18843-10G
OR18843-5G
OR18843-25G
APOSOR18843-10GEA
3910
CZK
APOSOR18843-10G
APOSOR18843-5G
APOSOR18843-25G
4-(Benzyloxy)cyclohexanone 96%
4-(Benzyloxy)cyclohexanone
Vzorec:
C₁₃H₁₆O₂ MW: 204,2700 g/mol Bod varu: 102…103 °C/0,15mmHg Storage Temperature: Ambient |
MDL Number:
MFCD10697867 CAS číslo: 2987-06-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...