Tisk…

3-Bromo-5-methylbenzaldehyde

Dodavatel: Apollo Scientific
Varování

OR18800-25G OR18800-5G OR18800-100G
APOSOR18800-25GEA 1620 CZK
APOSOR18800-25G APOSOR18800-5G APOSOR18800-100G
3-Bromo-5-methylbenzaldehyde
3-Bromo-5-methylbenzaldehyde
Vzorec: C8H7BrO
MW: 199,0446 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09836680
CAS číslo: 188813-04-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR