Benzyl methyl sulphone
Dodavatel: Apollo Scientific
OR1877-10G
OR1877-50G
APOSOR1877-10GEA
2070
CZK
APOSOR1877-10G
APOSOR1877-50G
Benzyl methyl sulphone
Benzyl methyl sulphone
Vzorec:
C₈H₁₀O₂S Bod tání: 124…127 °C Storage Temperature: Ambient |
MDL Number:
MFCD00025074 CAS číslo: 3112-90-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...