5-Bromo-4,6-dichloropyrimidine
Dodavatel: Apollo Scientific
|
Nebezpečí
|
OR1876-1G
OR1876-250MG
APOSOR1876-1GEA
705
CZK
APOSOR1876-1G
APOSOR1876-250MG
5-Bromo-4,6-dichloropyrimidine
5-Bromo-4,6-dichloropyrimidine
Vzorec:
C₄HBrCl₂N₂ MW: 227,88 g/mol Bod tání: 77…81 °C Storage Temperature: Ambient |
MDL Number:
MFCD00047379 CAS číslo: 68797-61-5 EINECS: 272-305-9 UN: 2923 ADR: 8+6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...