2-(Methoxycarbonylmethyl)phenylboronic acid pinacol ester 96%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
Methyl 2-(2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)acetate, 2-(2-Methoxy-2-oxoethyl)benzeneboronic acid, pinacol ester
OR18736-1G
OR18736-250MG
APOSOR18736-1GEA
8870
CZK
APOSOR18736-1G
APOSOR18736-250MG
2-(Methoxycarbonylmethyl)phenylboronic acid pinacol ester 96%
2-(Methoxycarbonylmethyl)phenylboronic acid pinacol ester
Vzorec:
C₁₅H₂₁BO₄ MW: 276,1358 g/mol Bod tání: 66…68 °C Storage Temperature: Ambient |
MDL Number:
MFCD12913983 CAS číslo: 956229-86-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...