Ethyl-4-methyl-1,3-thiazole-2-carboxylate
Dodavatel: Apollo Scientific
|
Varování
|
OR18721-1G
OR18721-250MG
APOSOR18721-1GEA
4570
CZK
APOSOR18721-1G
APOSOR18721-250MG
Ethyl-4-methyl-1,3-thiazole-2-carboxylate
Ethyl-4-methyl-1,3-thiazole-2-carboxylate
Vzorec:
C₇H₉NO₂S MW: 171,22 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD11112113 CAS číslo: 7210-73-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...