Tisk…

6-Chloro-1H-pyrrolo[2,3-b]pyridine-2-carboxylic acid 95%

Dodavatel: Apollo Scientific
Varování

OR18603-100MG OR18603-500MG
APOSOR18603-100MGEA 1120 CZK
APOSOR18603-100MG APOSOR18603-500MG
6-Chloro-1H-pyrrolo[2,3-b]pyridine-2-carboxylic acid 95%
6-Chloro-1H-pyrrolo[2,3-b]pyridine-2-carboxylic acid
Vzorec: C₈H₅ClN₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD09878690
CAS číslo: 800402-07-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR