Tisk…

Ethyl 2-amino-3-cyano-4,5,6,7-tetrahydro-1-benzothiophene-4-carboxylate

Dodavatel: Apollo Scientific
Varování

Synonyms: Ethyl 2-amino-3-cyano-4,5,6,7-tetrahydrobenzo[b]thiophene-4-carboxylate

OR18551-1G OR18551-500MG OR18551-5G
APOSOR18551-1GEA 6620 CZK
APOSOR18551-1G APOSOR18551-500MG APOSOR18551-5G
Ethyl 2-amino-3-cyano-4,5,6,7-tetrahydro-1-benzothiophene-4-carboxylate
Ethyl-2-amino-3-cyano-4,5,6,7-tetrahydro-1-benzothiophene-4-carboxylate
Vzorec: C₁₂H₁₄N₂O₂S
MW: 250.32 g/mol
Bod tání: 135…138 °C
Storage Temperature: Ambient
MDL Number: MFCD02679242
CAS číslo: 175599-83-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR