Pyrazine-2-carbaldehyde 97%
Dodavatel: Apollo Scientific
|
Varování
|
OR1854-1G
OR1854-250MG
OR1854-5G
APOSOR1854-1GEA
0
CZK
APOSOR1854-1G
APOSOR1854-250MG
APOSOR1854-5G
Pyrazine-2-carbaldehyde 97%
Pyrazine-2-carbaldehyde
Vzorec:
C5H4N2O Bod varu: 120…130 °C/30mmHg Storage Temperature: Refrigerator |
MDL Number:
MFCD02179278 CAS číslo: 5780-66-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...