Bromocyclopropane 99+%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
Cyclopropyl bromide
OR18407-100G
OR18407-25G
APOSOR18407-100GEA
0
CZK
APOSOR18407-100G
APOSOR18407-25G
Bromocyclopropane 99+%
Bromocyclopropane
Vzorec:
C₃H₅Br MW: 120,98 g/mol Bod varu: 69 °C Hustota: 1,51 g/cm³ (25 °C) Bod vzplanutí: –6 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00001271 CAS číslo: 4333-56-6 EINECS: 224-375-7 UN: 1993 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...