Tisk…

Methanesulphonamide 97%

Dodavatel: Apollo Scientific
Varování

Synonyms: Methylsulphonamide

OR18387-500G OR18387-100G OR18387-50G
APOSOR18387-500GEA 1780 CZK
APOSOR18387-500G APOSOR18387-100G APOSOR18387-50G
Methanesulphonamide 97%
Methanesulphonamide
Vzorec: CH₅NO₂S
MW: 95,13 g/mol
Bod tání: 85…93 °C
Storage Temperature: Ambient
MDL Number: MFCD00007940
CAS číslo: 3144-09-0
EINECS: 221-553-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR