Tisk…

Dimethyl-5-nitroisophthalate 98+%

Dodavatel: Apollo Scientific
Varování

Synonyms: Dimethyl 5-nitroisophthalate, 5-Nitroisophthalic acid dimethyl ester

OR18375-250G OR18375-100G OR18375-500G
APOSOR18375-250GEA 1080 CZK
APOSOR18375-250G APOSOR18375-100G APOSOR18375-500G
Dimethyl-5-nitroisophthalate 98+%
Dimethyl-5-nitroisophthalate
Vzorec: C₁₀H₉NO₆
MW: 239,18 g/mol
Bod tání: 121…125 °C
Storage Temperature: Ambient
MDL Number: MFCD00008429
CAS číslo: 13290-96-5
EINECS: 236-307-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR