Tisk…

Ethyl-3,4-dihydroxybenzoate

Dodavatel: Apollo Scientific
Varování

Synonyms: 3,4-Dihydroxybenzoic acid ethyl ester, Protocatechuic acid ethyl ester, Ethyl 3,4-dihydroxybenzoate

OR18368-25G OR18368-100G OR18368-500G
APOSOR18368-25GEA 622 CZK
APOSOR18368-25G APOSOR18368-100G APOSOR18368-500G
Ethyl-3,4-dihydroxybenzoate
Ethyl-3,4-dihydroxybenzoate
Vzorec: C₉H₁₀O₄
MW: 182,18 g/mol
Bod tání: 132…135 °C
Storage Temperature: Ambient
MDL Number: MFCD00002199
CAS číslo: 3943-89-3
EINECS: 223-529-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR