Tisk…

Ethyl-1-(3-amino-4-nitrophenyl)-4-piperidinecarboxylate

Dodavatel: Apollo Scientific

OR183520-1G OR183520-5G
APOSOR183520-1GEA 2440 CZK
APOSOR183520-1G APOSOR183520-5G
Ethyl-1-(3-amino-4-nitrophenyl)-4-piperidinecarboxylate
Ethyl-1-(3-amino-4-nitrophenyl)-4-piperidinecarboxylate
Vzorec: C₁₄H₁₉N₃O₄
MW: 293,33 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03425788
CAS číslo: 439095-45-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR