Fmoc-Osu (N-(9-fluorenylmethoxycarbonyloxy)succinimide)
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
Fmoc-OSu, N-(9-Fluorenylmethoxycarbonyloxy)succinimide
OR18352-100G
OR18352-25G
OR18352-1KG
APOSOR18352-100GEA
1240
CZK
APOSOR18352-100G
APOSOR18352-25G
APOSOR18352-1KG
Fmoc-Osu (N-(9-fluorenylmethoxycarbonyloxy)succinimide)
Fmoc-Osu (N-(9-fluorenylmethoxycarbonyloxy)succinimide)
Vzorec:
C₁₉H₁₅NO₅ MW: 337,3 g/mol Bod tání: 150…153 °C Storage Temperature: Ambient |
MDL Number:
MFCD00010733 CAS číslo: 82911-69-1 EINECS: 433-520-5 UN: 3077 ADR: 9+EHS,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...