Phthalaldehyde 98%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
1,2-Phthalic dicarboxaldehyde, Phthaldialdehyde, o-Phthalaldehyde, OPA, Phthaldicarboxaldehyde, Benzene-1,2-dicarboxaldehyde
OR18349-250G
OR18349-25G
OR18349-100G
APOSOR18349-250GEA
5200
CZK
APOSOR18349-250G
APOSOR18349-25G
APOSOR18349-100G
Phthalaldehyde 98%
Phthalaldehyde
Vzorec:
C₈H₆O₂ MW: 134,13 g/mol Bod varu: 83 °C/0,8mmHg Bod tání: 53…57 °C Hustota: 1,13 g/cm³ (25 °C) Bod vzplanutí: 132 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00003335 CAS číslo: 643-79-8 EINECS: 211-402-2 UN: 2923 ADR: 8+6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...