3-Nitrophthalic acid 96%
Dodavatel: Apollo Scientific
|
Varování
|
OR18338-1KG
OR18338-2.5KG
OR18338-500G
APOSOR18338-1KGEA
1870
CZK
APOSOR18338-1KG
APOSOR18338-2.5KG
APOSOR18338-500G
3-Nitrophthalic acid 96%
3-Nitrophthalic acid
Vzorec:
C₈H₅NO₆ Storage Temperature: Ambient |
MDL Number:
MFCD00007138 CAS číslo: 603-11-2 EINECS: 210-030-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...