Tisk…

Phenazine 99+%

Dodavatel: Apollo Scientific
Nebezpečí

OR18320-100G OR18320-25G OR18320-5G
APOSOR18320-100GEA 2780 CZK
APOSOR18320-100G APOSOR18320-25G APOSOR18320-5G
Phenazine 99+%
Phenazine
Vzorec: C₁₂H₈N₂
Bod tání: 172…177 °C
Storage Temperature: Ambient
MDL Number: MFCD00005023
CAS číslo: 92-82-0
EINECS: 202-193-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR