Tisk…

Diisopropylcarbodiimide 99%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: N,N'-methanetetraylbis(1-methylethylamine), N,N-Diisopropylcarbodiimide, 1,3-Diisopropylcarbodiimide, N,N'-Diisopropylcarbodiimide, N.N’-Diisopropylcarbodiimide, DIC

OR18304-100G OR18304-25G
APOSOR18304-100GEA 2280 CZK
APOSOR18304-100G APOSOR18304-25G
Diisopropylcarbodiimide 99%
Diisopropylcarbodiimide
Vzorec: C₇H₁₄N₂
MW: 126,2 g/mol
Bod varu: 145…148 °C
Hustota: 0,815 g/cm³ (25 °C)
Bod vzplanutí: 31 °C
Storage Temperature: Ambient
MDL Number: MFCD00065689
CAS číslo: 693-13-0
EINECS: 211-743-7
UN: 2929
ADR: 6.1+3,I

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR