2-Quinolinol 98%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
Quinolin-2(1H)-one, Quinolin-2-ol, Carbostyril, 2-Quinolone, 2-Hydroxyquinoline
OR18280-2.5G
OR18280-25G
OR18280-5G
APOSOR18280-2.5GEA
622
CZK
APOSOR18280-2.5G
APOSOR18280-25G
APOSOR18280-5G
2-Quinolinol 98%
2-Quinolinol
Vzorec:
C₉H₇NO MW: 145,16 g/mol Bod tání: 198…199 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006743 CAS číslo: 59-31-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...